Learn More
MyBioSource anti-COL5A1 antibody :: Rabbit anti-Human, mouse COL5A1 Polyclonal Antibody
SDP
Supplier:  MyBioSource MBS92094925X0.4 ML
Encompass
Species Reactivity- Human, mouseSpecificity-This COL5A1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 54-82 amino acids from the N-terminal region of human COL5A1.Purity/Purification- This antibody is purified through a protein A column, followed by peptide affinity purification.
Catalog No. NC3243042