Learn More
Cosmo Bio Usa Inc Anti-Cyclobutane Pyrimidine Dimers (CPDs) mAb (Clone TDM-2)
SDP
Supplier:  Cosmo Bio Usa Inc CACNMDND001
Encompass
Cyclobutane pyrimidine dimer (CPD)-specific mAb TDM-2 was cloned by fusing mouse myeloma cells with splenocytes from BALB/c mice immunized with methylated BSA conjugated with UVC-irradiated calf thymus DNA. TDM-2 binds CPDs in denatured DNA from all organisms from bacteria to human and is validated for ELISA and indirect immunofluorescence.
Catalog No. NC9036823