Learn More
Bio X Cell Monoclonal Antibody, Anti-Mouse CSF1, Rat IgG1, In Vivo Neutralization
Supplier: Bio X Cell BE0204R005MG
This monoclonal antibody, clone 5A1, reacts with mouse colony stimulating factor 1 (CSF1), also known as macrophage colony-stimulating factor (M-CSF). CSF1 is a hematopoietic growth factor expressed by various cell types including fibroblasts, stromal cells, osteoblasts, activated T cells, macrophages, and B cells. It regulates the proliferation, differentiation, and survival of monocytes, macrophages, and bone marrow progenitor cells. The 5A1 antibody has been demonstrated to neutralize CSF1 in vivo.
- Reacts with mouse colony stimulating factor 1 (CSF1)
- Neutralizes CSF1 in vivo
- Isotype is rat IgG1, κ
- Purity is greater than or equal to 95%, as determined by SDS-PAGE
- Formulated in PBS, pH 8.0, without stabilizers or preservatives
- Endotoxin levels are less than or equal to 1 EU/mg
- Filtered to 0.2 μm
- Produced in an animal-free facility
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.