Learn More
Selleck Chemical LLC Anti-CTLA-8-A2861-1MG
SDP
Supplier:  Selleck Chemical LLC A28611MG
Encompass_Preferred
Anti-CTLA-8 is a human monoclonal antibody against IL-17A (interleukin-17A) that can potently neutralize the effects of recombinant and native human IL-17A MW 143 72
Catalog No. 50-313-5313