Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-CYFRA21-1, 0.2mg
Supplier:  Advanced ImmunoChemical, Inc. 2CYFRA210.2MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal mouse anti-CYFRA21-1. MAbs:XC42. Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with soluble cytokeratin antigen fraction purified from human cell line. MAb Isotypes:IgG1 for MAb XC42. Applications:Immunoassays; (KS19.1 homology (high cross-inhibition)), immunoblotting.Purification:Ion exchange chromatographyPresentation:PBS, 0.05 % sodium azide (NaN3)Storage Conditions:+ 4 °CMaterial safety note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. 50-153-9869