Learn More
Seracare Life Sciences Inc ANTI-DEER IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 52200298
Encompass_Preferred
Affinity purified polyclonal antibody to Deer IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-669-87