Learn More
Metrohm USA Titration tip with antidiffusion valve / M6 thread
SDP
Supplier:  Metrohm USA 61543200
Encompass
Titration tip / M6 thread. Titration tip with antidiffusion valve. Length (mm) 151. Material ETFE/FEP. Parts: Antidiffusion valve For 6.1541.XXX and 6.1543.130, 6.1543.140, 6.1543.150, 6.1543.160, 6.1543.200 antidiffusion buret tips. 1 piece(s).
Catalog No. NC0022451