Learn More
DIMA BIOTECHNOLOGY LLC Anti-DLL3 antibody(67E6), IgG1 Chimeric mAb, 500 μg
Supplier:  DIMA BIOTECHNOLOGY LLC DMC101089500UG
Encompass
This gene encodes a member of the delta protein ligand family. This family functions as Notch ligands that are characterized by a DSL domain, EGF repeats, and a transmembrane domain. Mutations in this gene cause autosomal recessive spondylocostal dysostosis 1. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq, Jul 2008]
Catalog No. NC3353593