Learn More
BIOHIPPO Anti-DTYMK Polyclonal Antibody, 100 µg
SDP
Supplier:  BIOHIPPO BHA21404346
Encompass_Preferred
Human DTYMK Polyclonal Antibody is a Rabbit-derived polyclonal antibody that specifically recognizes DTYMK. It shows reactivity with Human, Mouse, Rat. Validated applications include ELISA, IHC, WB. Purified by antigen affinity column. Supplied as 100 ug.
Catalog No. 50-008-61034