Learn More
Sino Biological Anti-Dxd/Exatecan Antibody, Mouse Monoclonal
Supplier:  Sino Biological 68256MM06 100 UG
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Dxd conjugated to BSA. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. NC3853122