Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody Rabbit - Affinity Purified
SDP
Supplier:  Immunology Consultants RFLG45AZ
Encompass_Preferred
RFLG‑45A‑Z offers the same high‑quality affinity‑purified anti‑FLAG antibody as RFLG‑45A but in a 0.1 mg format ideal for small‑scale experiments, pilot studies, or limited‑use applications. This rabbit polyclonal antibody provides strong specificity for the DYKDDDDK epitope, delivering reliable detection in Western blotting, ELISA, and immunoprecipitation workflows. Produced by Immunology Consultants, RFLG‑45A‑Z is a cost‑efficient option for laboratories requiring a smaller quantity of a high‑performance anti‑FLAG reagent.
Catalog No. 50-004-35347