Learn More
Genscript Corporation Anti-DYKDDDDK G1 Affinity Resin, 25 ml settled resin, 50 ml total slurry, 4% cross-linked agarose, 90 μm average bead size, 1 mg binding capacity/ml
Supplier:  Genscript Corporation L0043225ML
Encompass
The GenScript Anti-DYKDDDDK G1 Affinity Resin is designed for the efficient purification of DYKDDDDK-tagged proteins from various expression systems such as bacteria, yeast, and mammalian cells. This affinity resin utilizes a high-affinity mouse monoclonal antibody coupled to 4% cross-linked agarose beads, allowing for specific protein capture while minimizing non-specific binding. It supports multiple elution methods and is ideal for both purification and immunoprecipitation applications.
  • High specificity and capacity for clean protein isolation
  • Versatile elution options for various protein characteristics
  • Compatible with bacteria, yeast, and mammalian cell systems
  • Reusable for up to 10 times with proper maintenance
Catalog No. NC2853910