Learn More
Abcam Rabbit Polyclonal EEA1 antibody Anti-EEA1
Supplier:  Abcam AB264245100UL
Encompass_Preferred
Rabbit Polyclonal EEA1 antibody. Suitable for IP, WB, IHC-P and reacts with Human samples.
Catalog No. NC3790586