Learn More
ECM Bioscience EphA2 (Extracellular region) M049 Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience EM0491
Encompass_Preferred
Clone (M049) was generated from a recombinant protein that included the extracellular region of human EphA2 protein. Accession #P29317. Clone M049 mouse monoclonal antibody detects a 125kDa* protein on SDS-PAGE "Native" immunoblots of human NCI-H2052 and MDA-MB-231 carcinomas, as well as the EphA2 C-terminal 60 kDa fragment in human A431 and A549. The antibody detects native EphA2, as well as aldehyde-fixed EphA2. The antibody works for Native western blot, immunoprecipitation, immunocytochemistry, and ELISA capture. Applications: WB(N), EC, IP, ICC; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = EphA2, P29317, Epithelial cell kinase, Tyrosine-protein kinase receptor ECK, Ephrin type-A receptor 2
Catalog No. 50-206-8702