Learn More
ECM Bioscience ERK1 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience EM2331
Encompass_Preferred
Clone (M233) was generated from a recombinant human ERK1 protein that included amino acids residues in the C-terminal region. This sequence is conserved in rat and mouse ERK1, and has low homology to ERK2, as well as other ERK family members. Accession #P27361. The antibody detects a 44 kDa* protein corresponding to ERK1 on SDS-PAGE immunoblots of human A431 epithelial cells. It does not detect ERK2. Applications: WB, E, ICC, IHC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = ERK, p42, p44, MAPK
Catalog No. 50-206-8707