Learn More
Haematologic Technologies Inc ANTI-FACTOR IX 100UG
Supplier:  Haematologic Technologies Inc AHIX5041
Encompass_Preferred
Antibody, Monoclonal, Anti-Human Factor IX, 100 micrograms, IgG1, Specificity: Human factor IX, human factor IXa, heavy chain of human factor IX and IXa, Properties: RIA (+), Blot (+), ELISA (+), IHC (+), 50% (v/v) glycerol/H2O, Store at -20 Celsius
Catalog No. 50-883-366