Learn More
Sigma Aldrich Fine Chemicals Biosciences ANTI-FLAG M1 Agarose Affinity Gel, 25 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A459625ML
Encompass
ANTI-FLAG M1 Agarose Affinity Gel is a purified mouse IgG2B monoclonal antibody covalently attached to agarose.
Catalog No. NC1910022
Only null left
Add to Cart
Request a Quote