Learn More
Raybiotech Inc Anti-GLP2 Antibody, Biotinylated, 50 µg
Supplier:  Raybiotech Inc 13010388B50
Encompass_Preferred
Rabbit anti-GLP2 Biotinylated Antibody, 50 ugThis antibody was produced from antiserum of rabbits immunized with a polypeptide immunogen. The IgG fraction was purified by Protein G/A affinity chromatography.
Catalog No. NC3792654