Learn More
Immunology Consultants anti-Glu-Glu Antibody (Rabbit) - Affinity Purified
SDP
Supplier:  Immunology Consultants RGLU45AZ
Encompass_Preferred
RGLU‑45A‑Z offers the same high‑performance affinity‑purified anti‑Glu‑Glu antibody as RGLU‑45A but in a 0.1 mg format ideal for small‑scale or pilot‑scale studies. This reagent provides strong, selective binding to the Glu‑Glu epitope and performs reliably in Western blotting, ELISA, and immunoprecipitation workflows. Produced by Immunology Consultants, RGLU‑45A‑Z is a cost‑effective option for laboratories requiring a smaller quantity of a high‑purity anti‑Glu‑Glu antibody.
Catalog No. 50-004-35467