Learn More
Seracare Life Sciences Inc ANTI-GOAT IGG (H+L) AB DL680
Supplier:  Seracare Life Sciences Inc 52300401
Encompass_Preferred
Affinity purified polyclonal antibody to goat IgG, both heavy and light chains (whole IgG) made in rabbit and labeled with DyLight 680. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-671-38