Learn More
Medchemexpress LLC Anti-GREM1 Gremlin A 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9904261MG
Encompass_Preferred
The Anti-GREM1/Gremlin Antibody is a humanized antibody expressed in CHO that targets GREM1/Gremlin The Anti-GREM1/Gremlin Antibody has a huIgG4SP heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for the Anti-GREM1/Gremlin Antibody can refer to Human IgG4 kappa Isotype Control (HY-P99003)
Catalog No. 50-003-25364