Learn More
Genscript Corporation Anti-GS Linker Affinity Resin
Supplier:  Genscript Corporation L0103610
Encompass
This product is designed for purification of proteins antibodies and compounds containing (GnS)m linker (n 2 m 2) from commonly used protein expression systems such as bacteria yeast and mammalian cells
Catalog No. 50-000-58399