Learn More
Seracare Life Sciences Inc ANTI-GUINEAPIG IGG(H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 52200366
Encompass_Preferred
Affinity purified polyclonal antibody to guinea pig IgG, both heavy and light chains (whole IgG) made in goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-28