Learn More
Bio X Cell InVivoMAb Anti-Human CD3, Clone OKT-3, Mouse IgG2a, 0.2 μm Filtered
Supplier: Bio X Cell BE00012R025MG
The InVivoMAb anti-human CD3 antibody, clone OKT-3, targets human CD3ε, a 20 kDa transmembrane protein integral to the TCR complex. Found on T lymphocytes, NK-T cells, and developing thymocytes, CD3ε is vital for TCR signaling, T cell activation, and antigen recognition. This antibody possesses in vivo immunosuppressive properties and effectively treats allograft rejection.
- Reacts with human CD3ε
- Used for in vitro T cell stimulation/activation
- Applicable for in vivo T cell depletion in humanized mice
- Facilitates ex vivo T cell inhibition for xenografts
- Suitable for flow cytometry
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level is ≤1EU/mg
- Purity is ≥95%
- Sterilized by 0.2 μm filtration
- Produced in an animal-free facility
- Purified using protein G
- Store at 4°C; do not freeze
- Has a molecular weight of 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.