Learn More
Bio X Cell Antibody, Anti-CCL2 (MCP-1), Mouse, Human, Rat, Clone 2H5
Supplier: Bio X Cell BE0185R005MG
This antibody reacts with mouse, human, and rat CCL2 (monocyte chemoattractant protein 1 or MCP-1), a key chemokine regulating the migration and infiltration of immune cells. CCL2 is implicated in various diseases with monocytic infiltrates. The antibody neutralizes CCL2's biological effects in vivo.
- Clone 2H5
- Reactivities: mouse, human, rat
- Isotype: Armenian Hamster IgG, κ
- Unconjugated
- Immunogen: CHO-expressed mouse MCP-1
- Applications: in vivo CCL2 neutralization, immunohistochemistry (frozen)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtered
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.