Learn More
Bio X Cell Antibody, anti-human/monkey CD28, Mouse IgG1, 5 mg
Supplier: Bio X Cell BE0291R005MG
This monoclonal antibody reacts with human and monkey CD28, a 45 kDa costimulatory receptor from the Ig superfamily, expressed by thymocytes, most peripheral T cells, and NK cells. It acts as a receptor for CD80 (B7-1) and CD86 (B7-2). Signaling through the antibody augments IL-2 and IL-2 receptor expression, and enhances cytotoxicity of CD3-activated T cells. It has demonstrated in vitro stimulation of human T cell proliferation.
- Isotype: Mouse IgG1, κ; Unconjugated
- Immunogen: Human CD28 transfected cell line
- Applications: In vitro T cell stimulation/activation, immunoprecipitation, flow cytometry, immunohistochemistry (frozen)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg; Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration; Production: Animal-free facility; Purification: Protein G
- Molecular weight: 150 kDa; Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.