Learn More
Genscript Corporation Anti-HA IP Resin
Supplier:  Genscript Corporation L007771
Encompass
Anti-HA IP Resin, with high specificity and affinity to HA tag, is ideal for quick and efficient immunoprecipitation of HA-tagged protein or HA-tagged protein-target protein complex.
Catalog No. NC2011088