Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Hemoglobin Human HBA10, 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 2HH100.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc.
Monoclonal Antibodies / Metabolic Syndrome /Monoclonal mouse anti-Hemoglobin human HbA10
MAbs: Hb4, Hb6 Hybridoma clones. Derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with glycated (in vitro) synthetic peptide corresponding to the N-terminal end of beta-globin
Specificity: Native human HbA10Mab
Isotypes: IgG1 for MAbs Hb4 and Hb6
Applications: HbA1c immunoassay
Purification: MAb 4 F12: Chromatography on protein A Sepharose and gel-filtration, other MAbs: Chromatography on protein A Sepharose
Presentation: PBS, pH 7.4, 0.1 % sodium azide (NaN3)
Storage Conditions: +4 °C
This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material. Contains sodium azide (0.1 %) as preservative.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.