Learn More
BIOGEMS INTERNATIONAL INC Anti-Human CD3 SAFIRE Size 100 g
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 0512125100
Encompass
The OKT3 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.
The OKT3 antibody is an immunosuppressive, which has proven to be an effective therapeutic agent in liver, heart, and renal allograft rejection.
Catalog No. 50-152-8952