Learn More
Vector Laboratories Polyclonal Rabbit anti‑Human CCNE1 / Cyclin E1 Antibody (Ile119, IHC, IF, WB) LS‑C175985, 100 µl
SDP
Supplier:  Vector Laboratories LSC175985100
Encompass
Cyclin E1 antibody LS-C175985 is an unconjugated rabbit polyclonal antibody to Cyclin E1 (CCNE1) (Ile119) from human. It is reactive with human, mouse and rat. Validated for IF, IHC and WB.
Catalog No. NC2758945