Learn More
Biolegend Brilliant Violet 421™ anti-human CD178 (Fas-L)
Supplier:  Biolegend 306412
Encompass_Preferred
Brilliant Violet 421™ anti-human CD178 (Fas-L) [NOK-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC; Size: 100 tests
Catalog No. NC1784760