Learn More
Mabtech Inc Anti-human CD28 mAb CD28-A, purified, 500 uL
Supplier:  Mabtech Inc 3608150
Encompass
This antibody can be used, together with antigen, to co-stimulate specific T cells. CD28-A can also be used together with anti-CD3 mAb for polyclonal activation of T cells.
Catalog No. NC1082719