Learn More
STEMCELL Technologies Anti-Human CD56 (NCAM) Antibody, Clone HCD56, APC, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60021AZ.1
Encompass
Clone HCD56 is a mouse monoclonal antibody that reacts with human CD56 (NCAM), a member of the immunoglobulin (Ig) super family containing 5 Ig-like domains and two fibronectin type-3 domains in its extracellular portion. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD56 (NCAM). The ~140 kDa isoform of CD56 is considered the prototypic cell surface marker on human NK cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Buffy Coat CD56 Positive Selection Kit (Catalog #18085HLA) and EasySep™ Human CD56 Positive Selection Kit (Catalog #18055); partial blocking may be observed, as well as EasySep™ HLA CD2 Positive Selection Kit (Catalog #18657HLA) and EasySep™ Human NK Cell Enrichment Kit (Catalog #19055). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5077