Learn More
Bio X Cell InVivoSIM Anti-Human Claudin 18.2 (Zolbetuximab Biosimilar), Unconjugated Human IgG1 κ, 5 mg (SIM0026-5MG)
Supplier: Bio X Cell SIM00265MG
InVivoSIM anti-human Claudin 18.2 (Zolbetuximab biosimilar) is a research-grade, unconjugated monoclonal antibody (human IgG1 κ) that recognizes isoform 2 of human claudin 18 (CLDN18.2). It is supplied in PBS (pH 7.0) and intended for non-therapeutic research applications, including functional assays.
- Unconjugated human IgG1, κ monoclonal antibody.
- Reactivity with human Claudin 18.2 (isoform 2).
- Formulated in PBS, pH 7.0, without preservatives.
- Purity ≥95% (SDS-PAGE).
- Endotoxin ≤0.5 EU/mg.
- Aggregation <5% (SEC).
- Sterile-filtered through 0.2 μm.
- Produced in an animal-free facility and purified by Protein A.
- Store at 4°C; freezing is not recommended.
- Reported applications: functional assays and other research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.