Learn More
Immunology Consultants Monoclonal Mouse anti-Human Epididymis Protein 4 (HE4) Antibody Clone# 3A8, 1.0 mg
SDP
Supplier:  Immunology Consultants MHE480A3A8
Encompass_Preferred
This mouse IgG1 kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Human Epididymis Protein 4 (HE4) as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator
Catalog No. NC2734218