Learn More
BIOHIPPO Anti-Human FLT4/VEGFR3 Antibody (SAA2200), PerCP, 100 Tests
SDP
Supplier:  BIOHIPPO BHA21419987
Encompass_Preferred
Human FLT4/VEGFR3 Antibody (clone SAA2200) is a PerCP-conjugated monoclonal antibody for flow cytometry and related applications. It detects FLT4/VEGFR3 in Human samples. Validated applications include ELISA, FCM. Supplied as 100 tests.
Catalog No. 50-008-57141