Learn More
Mabtech Inc ANTI-HUMAN IFN GAMMA 1D1K 1MG
Supplier:  Mabtech Inc 342031000
Encompass
anti-human IFN-γ mAb 1-D1K, purified. 1 MG. Monoclonal antibody specific for IFN-γ. This monoclonal antibody enables sensitive and specific detection of human IFN-γ in immunoassays such as ELISA and ELISpot. We recommend 1-D1K as capture antibody in ELISA and ELISpot in combination with biotinylated detection antibody 7-B6-1. CONTENTS: 1-D1K, purified, supplied at 1 mg/ml in PBS with 0.02% sodium azide.
Catalog No. NC9586493