Learn More
REVMAB BIOSCIENCES USA INC Ig Light Chain Rabbit Anti-Human Monoclonal Antibody [RM129]
Supplier:  REVMAB BIOSCIENCES USA INC 32103100
Encompass_Preferred
Anti-Human Ig Light Chain Rabbit Monoclonal Antibody, Clone RM129 (100ug); ICC, IHC, FC, ELISA validated; Product info: www.revmab.com
Catalog No. NC1494180