Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (FC) ANTIBODY
Supplier:  Seracare Life Sciences Inc 52100165
Encompass_Preferred
Affinity purified polyclonal secondary antibody against the Fc fragment of Human IgG (whole IgG) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-09