Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (G) ANTIBODY
Supplier:  Seracare Life Sciences Inc 52100155
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-03