Learn More
Immunology Consultants HRP Conjugated Goat anti-Human IgM Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants GM80P
Encompass_Preferred
Goats were immunized with highly purified human IgM and the resulting antiserum was collected. Antibody was solid phase adsorbed, immunoaffinity purified off a human IgM containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG.This antibody reacts with human IgM as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may cross-react with IgM from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4288