Learn More
Rockland Immunochemicals Human IgM (mu chain) Antibody Peroxidase Conjugated 1 mg
SDP
Supplier:  Rockland Immunochemicals 6091307
Encompass_Preferred
Anti-Human IgM (mu heavy chain) Peroxidase generated in goat detects specifically Human IgM mu heavy chain. Immunoglobulin M is the largest antibody isotype and the first to be secreted against an initial exposure to antigen. IgM is predominantly produced in the spleen. Formed from covalently linking 5 immunoglobulins together. Anti-Human IgM mu chain antibody is ideal for investigators in Immunology Microbiology and Cell Biology.
Catalog No. 50-194-3889