Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Human Leptin, 0.2mg
Supplier:  Advanced ImmunoChemical, Inc. 1L10.2MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal mouse anti-human leptin. MAbs:3G7, 4F12Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant leptin.Specificity:MAbs recognize recombinant human leptin and leptin circulating in human blood. MAb Isotypes:IgG1 for all MAbs for all MAbsApplications:Leptin immunoassay. All MAbs recognize human leptin in Western blotting. MAbs were tested in sandwich immunoassay with human blood samples. All recommended pairs recognize antigen in human blood with high sensitivity. Recommended pairs for sandwich immunoassay (capture – detection): 3G7 – 4F124F12 – 3G7. Purification: MAb 4F12: Chromatography on protein A Sepharose and gel-filtration, other MAbs: Chromatography on protein A Sepharose. Presentation: PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C.
Catalog No. 50-153-9864