Learn More
FUJIFILM BIOSCIENCES INC Anti-Human Macrophage Scavenge
Supplier:  FUJIFILM BIOSCIENCES INC 30409751
Encompass_Preferred
This antibody is designed for research use in detecting human macrophage scavenger receptor CD204 aiding in studies related to immune response and macrophage activity Specificity for human CD204 validated for IH and WB
Catalog No. 50-004-50127