Learn More
BIOHIPPO Anti-Human MET/c-Met/HGFR Antibody (SAA0111), 100 µg
SDP
Supplier:  BIOHIPPO BHA21413190
Encompass_Preferred
Human MET/c-Met/HGFR Antibody (clone SAA0111) is a monoclonal antibody for flow cytometry and related applications. It detects MET/c-Met/HGFR in Human samples. Validated applications include FCM. Supplied as 100 ug.
Catalog No. 50-008-59079