Learn More
STEMCELL Technologies Anti-Human MUC1 (CD227) Antibody, Clone 214D4, Size: 200 µg
SDP
Supplier:  STEMCELL Technologies 60137
Encompass
Clone 214D4 is a mouse monoclonal antibody that reacts with human MUC1 (CD227), a large cell surface glycoprotein expressed by most glandular and ductal epithelial cells as well as a variety of hematopoietic cells. Verified for use with flow cytometry and immunohistochemistry assays, use this antibody in applications requiring the detection of MUC1 (CD227). A characteristic feature of the MUC1 (CD227) glycoprotein is a core protein domain composed of a variable number of tandem repeats and multiple oligosaccharide side chains. The protein is highly expressed by the majority of human adenocarcinomas and is associated with a poor prognosis. The 214D4 antibody clone is particularly useful since it enriches for luminal-restricted but not bipotent epithelial progenitor cells from normal human mammary tissue. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5713