Learn More
FUJIFILM BIOSCIENCES INC Anti Human Podocalyxin Monoclo
Supplier:  FUJIFILM BIOSCIENCES INC 01127491
Encompass_Preferred
Podocalyxin is a member of the CD34 family of type I transmembrane sialomucins It is expressed in many normal cells such as renal podocytes and endothelial cells It is also expressed in many types of tumors including colorectal cancers breast cancers and brain tumors Overexpression of podocalyxin is an independent predictor of progression metastasis and poor prognosis Recent studies indicate that the glycosylation of podocalyxin change depending on the state of the cells This antibody recognized endogenous podocalyxin expressing cancer cell lines and normal cells independently of glycosylation
Catalog No. 50-004-50168