Learn More
BIOHIPPO Anti-Human TERT Antibody (SAA1735), 100 µg
SDP
Supplier:  BIOHIPPO BHA21407237
Encompass_Preferred
TERT Recombinant Antibody (clone SAA1735) is a recombinant Mouse IgG2a, kappa antibody specific for TERT. It reacts with Human. Suitable for ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60175