Learn More
BIOHIPPO Anti-IARS1 Polyclonal Antibody, 100 µg
SDP
Supplier:  BIOHIPPO BHA21400571
Encompass_Preferred
Human IARS1 Polyclonal Antibody is a Rabbit-derived polyclonal antibody that specifically recognizes IARS1. It shows reactivity with Human, Mouse, Rat. Validated applications include ELISA, IHC, WB. Purified by antigen affinity column. Supplied as 100 ug.
Catalog No. 50-008-59547