Learn More
MOLECULAR DEPOT LLC Anti-IgM Latex - 1 mL
Supplier:  MOLECULAR DEPOT LLC B2010396
Encompass_Preferred
Anti-IgM Latex Particles is a suspension of latex particles (0 35 um 0 5perc solid) conjugated to polyclonal anti human IgM antibodies Highly specific and highly stable these particles can be used for the easy one-step capture of various antigens proteins and antibodies specific to human IgM antibodies These particles can be recovered easily by centrifugation allowing for various applications such as purification depletion assay development and more For in vitro research use only
Catalog No. 50-000-13359